Gene Transcripts
- Gene Symbol
- 3110039I08Rik
- Entrez Gene ID
- 73144
- Gene Name
- RIKEN cDNA 3110039I08 gene
- Gene Aliases
- 2610203C20Rik, AI848448, D230004N17Rik, D3Mgi9, Mir100hg
- Chromosome Location
- Chr. 9: 41376561 - 41432079 on Build GRCm38
- UniGene ID
- Mm.38166
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK034202.1 |
- |
1 |
3185 |
|
AK034222.1 |
- |
2 |
1769 |
|
AK141402.1 |
- |
|
2122 |
|
View Full Product Details