Gene Transcripts

Gene Symbol
AW146154
Entrez Gene ID
101835
Gene Name
expressed sequence AW146154
Gene Aliases
AI451288
Chromosome Location
Chr. 7: 41478874 - 41499890 on Build GRCm38
UniGene ID
Mm.370336
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033530.3 NP_001028702.1 1 176
GenBank mRNA AK164198.1 - 1 177

View Full Product Details