Gene Transcripts

Gene Symbol
Calr4
Entrez Gene ID
108802
Gene Name
calreticulin 4
Gene Aliases
4933403L16Rik, AU017583
Chromosome Location
Chr. 4: 109204779 - 109255138 on Build GRCm38
UniGene ID
Mm.318248
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033226.4 NP_001028398.1 1 63
GenBank mRNA AK163350.1 - 1 64

View Full Product Details