Gene Transcripts
- Gene Symbol
- Slc19a1
- Entrez Gene ID
- 29723
- Gene Name
- solute carrier family 19 (folate transporter), member 1
- Gene Aliases
- MTX1
- Chromosome Location
- Chr. 20: 12334675 - 12353853 on Build Rnor_6.0
- UniGene ID
- Rn.9042
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001035232.1 |
NP_001030309.1
|
2, 3 |
306 |
|
NM_017299.2 |
NP_058995.2
|
2, 3 |
439 |
|
XM_006256295.2 |
XP_006256357.1
|
2, 3 |
1328 |
|
XM_006256296.2 |
XP_006256358.1
|
2, 3 |
331 |
|
XM_006256297.2 |
XP_006256359.1
|
2, 3 |
755 |
|
XM_006256298.2 |
XP_006256360.1
|
2, 3 |
317 |
|
GenBank mRNA |
AF099009.1 |
- |
2, 3 |
448 |
|
AF099010.1 |
- |
2, 3 |
448 |
|
AF173642.1 |
- |
2, 3 |
285 |
|
AY756271.1 |
- |
2, 3 |
285 |
|
AY756272.1 |
- |
2, 3 |
285 |
|
AY756273.1 |
- |
2, 3 |
285 |
|
BC085742.1 |
- |
2, 3 |
306 |
7193913 |
U38180.1 |
- |
2, 3 |
431 |
|
View Full Product Details