Gene Transcripts

Gene Symbol
Olfr1274-ps
Entrez Gene ID
258330
Gene Name
olfactory receptor 1274, pseudogene
Gene Aliases
GA_x5J8B7W2BV0-3116-4045, MOR228-4, MOR228-5, Olfr1274
Chromosome Location
Chr. 2: 90400640 - 90401674 on Build GRCm38
UniGene ID
Mm.330392
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146263.2 NP_666375.2 1 423
GenBank mRNA BC145937.1 - 423

View Full Product Details