Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017313.3 | NP_059009.2 | 2 | 518 | |
XM_006241357.2 | XP_006241419.1 | 3 | 687 | ||
XM_006241358.2 | XP_006241420.1 | 3 | 692 | ||
XM_006241359.2 | XP_006241421.1 | 3 | 1696 | ||
XM_006241360.2 | XP_006241422.1 | 3 | 682 | ||
XM_008765382.1 | XP_008763604.1 | 2 | 744 | ||
XM_008765383.1 | XP_008763605.1 | 1 | 57 | ||
GenBank mRNA | BC127500.1 | - | 2 | 521 | |
U19181.1 | - | 2 | 511 |