Gene Transcripts

Gene Symbol
Cecr2
Entrez Gene ID
330409
Gene Name
cat eye syndrome chromosome region, candidate 2
Gene Aliases
2610101O16Rik, 2810409N01Rik, Gtl4, mKIAA1740
Chromosome Location
Chr. 6: 120666320 - 120771194 on Build GRCm38
UniGene ID
Mm.333357
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128151.1 NP_001121623.1 17 3784
GenBank mRNA AK129435.1 - 6 2282
BC060152.1 - 1 148

View Full Product Details