Gene Transcripts
- Gene Symbol
- Gm10033
- Entrez Gene ID
- 378466
- Gene Name
- predicted gene 10033
- Gene Aliases
- 9330175B10Rik, ENSMUSG00000057924, ENSMUSG00000071083, Gm10311
- Chromosome Location
- Chr. 8: 69371025 - 69395544 on Build GRCm38
- UniGene ID
- Mm.37846
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK034302.1 |
- |
6 |
1072 |
|
AK129029.1 |
- |
6 |
1994 |
|
View Full Product Details