Gene Transcripts
- Gene Symbol
- Tnfrsf8
- Entrez Gene ID
- 25069
- Gene Name
- tumor necrosis factor receptor superfamily, member 8
- Gene Aliases
- Cd30
- Chromosome Location
- Chr. 5: 163186349 - 163232139 on Build Rnor_6.0
- UniGene ID
- Rn.11322
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019135.1 |
NP_062008.1
|
12, 13 |
1321 |
|
GenBank mRNA |
D42117.1 |
- |
12, 13 |
1321 |
|
View Full Product Details