Gene Transcripts

Gene Symbol
Zfp580
Entrez Gene ID
68992
Gene Name
zinc finger protein 580
Gene Aliases
1500015O20Rik, AI838225, Znf580
Chromosome Location
Chr. 7: 5051532 - 5053723 on Build GRCm38
UniGene ID
Mm.41484
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026900.1 NP_081176.1 2 833
GenBank mRNA AK005257.1 - 2 828
BC066001.1 - 2 781 5700640

View Full Product Details