Gene Transcripts

Gene Symbol
2310009B15Rik
Entrez Gene ID
69549
Gene Name
RIKEN cDNA 2310009B15 gene
Gene Aliases
Chromosome Location
Chr. 1: 138851979 - 138856854 on Build GRCm38
UniGene ID
Mm.264673
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081226.1 NP_001074695.1 3 429
GenBank mRNA AK009246.1 - 3 602

View Full Product Details