Gene Transcripts

Gene Symbol
Lif
Entrez Gene ID
60584
Gene Name
leukemia inhibitory factor
Gene Aliases
Chromosome Location
Chr. 14: 84492909 - 84500550 on Build Rnor_6.0
UniGene ID
Rn.44379
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022196.2 NP_071532.2 3 547
XM_006251364.2 XP_006251426.1 4 682
XM_008770359.1 XP_008768581.1 3 1252
GenBank mRNA AB010275.1 - 3 547
M32748.1 - 3 547

View Full Product Details