Gene Transcripts

Gene Symbol
Clic4
Entrez Gene ID
83718
Gene Name
chloride intracellular channel 4
Gene Aliases
Chromosome Location
Chr. 5: 153568937 - 153625669 on Build Rnor_6.0
UniGene ID
Rn.104756
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031818.1 NP_114006.1 2 127
GenBank mRNA AF104119.1 - 2 127

View Full Product Details