Gene Transcripts

Gene Symbol
Ralbp1
Entrez Gene ID
84014
Gene Name
ralA binding protein 1
Gene Aliases
Chromosome Location
Chr. 9: 113579107 - 113598477 on Build Rnor_6.0
UniGene ID
Rn.7107
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032067.1 NP_114456.1 1, 2 361
GenBank mRNA BC089788.1 - 1, 2 415
U28830.1 - 1, 2 365
U82623.1 - 1, 2 361

Gene Symbol
LOC501456
Entrez Gene ID
501456
Gene Name
similar to GTPase activating protein testicular GAP1
Gene Aliases
Chromosome Location
Chr. 16: 514156 - 518420 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008771014.1 XP_008769236.1 2 425

Gene Symbol
LOC680627
Entrez Gene ID
680627
Gene Name
similar to GTPase activating protein testicular GAP1
Gene Aliases
Chromosome Location
Chr. 16: 513812 - 622377 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008771013.1 XP_008769235.1 14 2272

View Full Product Details