Gene Transcripts

Gene Symbol
Cysltr1
Entrez Gene ID
114099
Gene Name
cysteinyl leukotriene receptor 1
Gene Aliases
Cyslt1
Chromosome Location
Chr. X: 77671028 - 77700491 on Build Rnor_6.0
UniGene ID
Rn.74253
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053641.1 NP_446093.1 1 86
XM_006257158.2 XP_006257220.1 3 800
GenBank mRNA AB052685.1 - 1 86

View Full Product Details