Gene Transcripts

Gene Symbol
Nox1
Entrez Gene ID
114243
Gene Name
NADPH oxidase 1
Gene Aliases
Chromosome Location
Chr. X: 104909328 - 104932508 on Build Rnor_6.0
UniGene ID
Rn.220465
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053683.1 NP_446135.1 9 1173
GenBank mRNA AF152963.1 - 9 1173

View Full Product Details