Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014906.4 | NP_055721.3 | 2 | 692 | |
NR_048561.1 | 2 | 692 | |||
XM_005257169.4 | XP_005257226.1 | 2 | 117 | ||
XM_011524534.2 | XP_011522836.1 | 2 | 1013 | ||
XM_017024359.1 | XP_016879848.1 | 2 | 1209 | ||
GenBank mRNA | AB028995.1 | - | 2 | 696 | |
AF260269.1 | - | 2 | 723 | ||
AF520614.1 | - | 2 | 569 | ||
AK289966.1 | - | 2 | 698 | ||
BC136290.1 | - | 2 | 638 | 9020277 | |
BC143845.1 | - | 2 | 638 | ||
BC151228.1 | - | 2 | 696 | 9007247 | |
CR749253.1 | - | 2 | 132 |