Gene Transcripts
- Gene Symbol
- Slc8a3
- Entrez Gene ID
- 140448
- Gene Name
- solute carrier family 8 (sodium/calcium exchanger), member 3
- Gene Aliases
- Ncx3
- Chromosome Location
- Chr. 6: 104891378 - 105097054 on Build Rnor_6.0
- UniGene ID
- Rn.67030
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_078620.1 |
NP_511175.1
|
2 |
1068 |
|
GenBank mRNA |
U53420.1 |
- |
2 |
1068 |
|
View Full Product Details