Gene Transcripts

Gene Symbol
Lcp2
Entrez Gene ID
155918
Gene Name
lymphocyte cytosolic protein 2
Gene Aliases
Slp76
Chromosome Location
Chr. 10: 18996523 - 19043289 on Build Rnor_6.0
UniGene ID
Rn.1057
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130421.2 NP_569105.1 1, 2 242
GenBank mRNA AB072980.1 - 1, 2 82
BC100066.1 - 1, 2 242 7387128
FQ232596.1 - 1, 2 256

View Full Product Details