Gene Transcripts

Gene Symbol
Zfp483
Entrez Gene ID
170955
Gene Name
zinc finger protein 483
Gene Aliases
HIT-10, Hit10, Zkscan16, Znf483
Chromosome Location
Chr. 5: 76090015 - 76103051 on Build Rnor_6.0
UniGene ID
Rn.211991
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133422.1 NP_596913.1 6 1624
GenBank mRNA AF277901.1 - 6 1624

View Full Product Details