Gene Transcripts

Gene Symbol
Mpc1
Entrez Gene ID
171087
Gene Name
mitochondrial pyruvate carrier 1
Gene Aliases
Brp44l
Chromosome Location
Chr. 1: 53026608 - 53038229 on Build Rnor_6.0
UniGene ID
Rn.3718
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133561.1 NP_598245.1 4 353
GenBank mRNA AF304429.1 - 4 353
BC097287.1 - 4 369 7445460
FQ219877.1 - 4 407

View Full Product Details