Gene Transcripts

Gene Symbol
Ppif
Entrez Gene ID
282819
Gene Name
peptidylprolyl isomerase F
Gene Aliases
CyP-D, CypD, PPIase
Chromosome Location
Chr. 16: 1979191 - 1985912 on Build Rnor_6.0
UniGene ID
Rn.2923
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172243.1 NP_758443.1 4 383
GenBank mRNA BC086977.1 - 4 416
U68544.1 - 4 383

View Full Product Details