Gene Transcripts

Gene Symbol
Rpl29
Entrez Gene ID
19944
Gene Name
ribosomal protein L29
Gene Aliases
Rpl43
Chromosome Location
Chr. 9: 106429432 - 106431568 on Build GRCm38
UniGene ID
Mm.288696
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009082.2 NP_033108.1 4 291
GenBank mRNA AK013307.1 - 4 291
AK088551.1 - 4 230
AK141477.1 - 4 256
AK167284.1 - 4 233
BC002062.1 - 209 3590425
BC019179.1 - 220
BC081467.1 - 3 203 6823037
BC082292.1 - 4 209 6487045
BC086897.1 - 4 237 6775819
BC086898.1 - 4 233 30120979
BC087950.1 - 4 233 30122718
BC092262.1 - 4 219 2651191
BC094039.1 - 4 219 5006561
BC096575.1 - 4 226 1430129
L08651.1 - 4 209

Gene Symbol
4931440P22Rik
Entrez Gene ID
71004
Gene Name
RIKEN cDNA 4931440P22 gene
Gene Aliases
Chromosome Location
Chr. 3: 65527483 - 65529414 on Build GRCm38
UniGene ID
Mm.475465
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK161073.1 - 707

View Full Product Details