Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009429.3 | NP_033455.1 | 3 | 362 | |
GenBank mRNA | AK149695.1 | - | 3 | 288 | |
AK150300.1 | - | 3 | 289 | ||
AK150500.1 | - | 3 | 288 | ||
AK150858.1 | - | 3 | 289 | ||
AK151059.1 | - | 3 | 290 | ||
AK151266.1 | - | 3 | 288 | ||
AK151375.1 | - | 3 | 288 | ||
AK151894.1 | - | 3 | 288 | ||
AK152690.1 | - | 3 | 288 | ||
AK152808.1 | - | 3 | 289 | ||
AK159516.1 | - | 3 | 288 | ||
AK159565.1 | - | 3 | 288 | ||
AK165736.1 | - | 3 | 288 | ||
AK166584.1 | - | 3 | 291 | ||
AK167413.1 | - | 3 | 288 | ||
AK167936.1 | - | 3 | 288 | ||
AK168151.1 | - | 3 | 288 | ||
AK169730.1 | - | 3 | 290 | ||
BC092381.1 | - | 3 | 248 | 6824238 | |
X06407.1 | - | 3 | 289 |