Gene Transcripts
- Gene Symbol
- Zfpm1
- Entrez Gene ID
- 22761
- Gene Name
- zinc finger protein, multitype 1
- Gene Aliases
- FOG, Fog1
- Chromosome Location
- Chr. 8: 122282141 - 122337862 on Build GRCm38
- UniGene ID
- Mm.3105
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_009569.3 |
NP_033595.1
|
10 |
3360 |
|
GenBank mRNA |
AF006492.1 |
- |
10 |
3364 |
|
View Full Product Details