Gene Transcripts
- Gene Symbol
- CDC25B
- Entrez Gene ID
- 994
- Gene Name
- cell division cycle 25B
- Gene Aliases
- Chromosome Location
- Chr. 20: 3786772 - 3806121 on Build GRCh38
- UniGene ID
- Hs.153752
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001287516.1 |
NP_001274445.1
|
8, 9 |
1016 |
|
NM_001287517.1 |
NP_001274446.1
|
8, 9 |
974 |
|
NM_001287519.1 |
NP_001274448.1
|
8, 9 |
747 |
|
NM_001287520.1 |
NP_001274449.1
|
8, 9 |
789 |
|
NM_001287522.1 |
NP_001274451.1
|
7, 8 |
666 |
|
NM_001287524.1 |
NP_001274453.1
|
4, 5 |
571 |
|
NM_004358.4 |
NP_004349.1
|
8, 9 |
1599 |
|
NM_021872.3 |
NP_068658.1
|
7, 8 |
1518 |
|
NM_021873.3 |
NP_068659.1
|
8, 9 |
1641 |
|
NR_136335.1 |
|
8, 9 |
789 |
|
NR_136336.1 |
|
8, 9 |
1016 |
|
GenBank mRNA |
AK092713.1 |
- |
8, 9 |
789 |
|
AK295573.1 |
- |
8, 9 |
858 |
|
AK299192.1 |
- |
8, 9 |
792 |
|
BC006395.2 |
- |
8, 9 |
922 |
4135465 |
BC009953.2 |
- |
8, 9 |
1002 |
4130804 |
BC051711.1 |
- |
8, 9 |
1546 |
5301205 |
BX640836.1 |
- |
4, 5 |
563 |
|
BX647988.1 |
- |
8, 9 |
704 |
|
FR695900.1 |
- |
8, 9 |
789 |
|
FR695901.1 |
- |
7, 8 |
666 |
|
M81934.1 |
- |
8, 9 |
878 |
|
S78187.1 |
- |
8, 9 |
1046 |
|
Z68092.1 |
- |
7, 8 |
826 |
|
View Full Product Details