Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001161769.1 | NP_001155241.1 | 5 | 1118 | |
NM_001161770.1 | NP_001155242.1 | 4 | 1250 | ||
NM_010723.3 | NP_034853.1 | 5 | 1405 | ||
GenBank mRNA | AF102817.1 | - | 5 | 1034 | |
AK080534.1 | - | 5 | 1055 | ||
AK138278.1 | - | 4 | 1251 | ||
AK159590.1 | - | 5 | 1332 | ||
BC003488.1 | - | 4 | 1117 | 2811636 | |
BC004661.1 | - | 5 | 1058 | ||
BC010278.1 | - | 5 | 1148 | 3589532 | |
BC100382.1 | - | 5 | 1010 | 6487478 |