Gene Transcripts

Gene Symbol
Mdk
Entrez Gene ID
17242
Gene Name
midkine
Gene Aliases
MK, Mek
Chromosome Location
Chr. 2: 91929805 - 91932297 on Build GRCm38
UniGene ID
Mm.906
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012335.2 NP_001012335.1 5 590
NM_001012336.2 NP_001012336.1 5 587
NM_001291481.1 NP_001278410.1 5 497
NM_001291482.1 NP_001278411.1 4 510
NM_001291483.1 NP_001278412.1 4 627
NM_010784.5 NP_034914.1 5 789
GenBank mRNA AK032742.1 - 5 493
AK084457.1 - 5 510
AY864926.1 - 5 523
BC012244.1 - 5 458 4167496
CF728526.1 - 5 483
CF738400.1 - 5 595
CF740555.1 - 3 307
CJ174578.1 - 4 385
DQ323886.1 - 5 565
DQ323887.1 - 4 598
M19662.1 - 4 668
M34327.1 - 5 463
M34328.1 - 5 774
M35833.1 - 5 463

View Full Product Details