Gene Transcripts

Gene Symbol
Stat3
Entrez Gene ID
20848
Gene Name
signal transducer and activator of transcription 3
Gene Aliases
1110034C02Rik, AW109958, Aprf
Chromosome Location
Chr. 11: 100886810 - 100939570 on Build GRCm38
UniGene ID
Mm.249934
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011486.4 NP_035616.1 5, 6 755
NM_213659.2 NP_998824.1 5, 6 755
NM_213660.2 NP_998825.1 5, 6 755
GenBank mRNA AK145486.1 - 5, 6 611
AK153005.1 - 5, 6 667
AK153472.1 - 5, 6 667
AY299489.1 - 4, 5 482
AY299490.1 - 4, 5 482
BC003806.1 - 5, 6 643 3593588
BC019168.1 - 5, 6 659 4923137
L29278.1 - 5, 6 734
U06922.1 - 5, 6 544
U08378.1 - 5, 6 622
U30709.1 - 5, 6 648

View Full Product Details