Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290707.1 | NP_001277636.1 | 4, 5 | 601 | |
NM_011866.2 | NP_035996.2 | 3, 4 | 555 | ||
GenBank mRNA | AF110507.1 | - | 3, 4 | 426 | |
AK039249.1 | - | 4, 5 | 601 | ||
AK147690.1 | - | 3, 4 | 522 | ||
AK162221.1 | - | 3, 4 | 209 | ||
AY360383.1 | - | 3, 4 | 555 | ||
BC113201.1 | - | 3, 4 | 421 | 40048238 |