Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013566.2 | NP_038594.2 | 4 | 591 | |
GenBank mRNA | AK078871.1 | - | 4 | 592 | |
AK144773.1 | - | 4 | 563 | ||
AK154766.1 | - | 4 | 592 | ||
AK155207.1 | - | 4 | 558 | ||
AK155594.1 | - | 4 | 563 | ||
BC011184.1 | - | 4 | 564 | 4187781 | |
M68903.1 | - | 4 | 539 | ||
M95632.1 | - | 4 | 535 | ||
M95633.1 | - | 4 | 535 | ||
S44607.1 | - | 3 | 503 |