Gene Transcripts
- Gene Symbol
- Mbd3
- Entrez Gene ID
- 17192
- Gene Name
- methyl-CpG binding domain protein 3
- Gene Aliases
- AI181826, AU019209
- Chromosome Location
- Chr. 10: 80392539 - 80400702 on Build GRCm38
- UniGene ID
- Mm.7142
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF072248.1 |
- |
1 |
348 |
|
CX208033.1 |
- |
1 |
240 |
|
View Full Product Details