Gene Transcripts

Gene Symbol
Cntn6
Entrez Gene ID
53870
Gene Name
contactin 6
Gene Aliases
NB-3
Chromosome Location
Chr. 6: 104492704 - 104863406 on Build GRCm38
UniGene ID
Mm.321671
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017383.3 NP_059079.2 3 412
GenBank mRNA AB032602.1 - 2 98
AK052972.1 - 4 305
BC076594.1 - 3 412 30649661

View Full Product Details