Gene Transcripts
- Gene Symbol
- MAK
- Entrez Gene ID
- 4117
- Gene Name
- male germ cell associated kinase
- Gene Aliases
- RP62
- Chromosome Location
- Chr. 6: 10762723 - 10838954 on Build GRCh38
- UniGene ID
- Hs.446125
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001242385.1 |
NP_001229314.1
|
2, 3 |
412 |
|
NM_001242957.2 |
NP_001229886.1
|
1, 2 |
338 |
|
NM_005906.5 |
NP_005897.1
|
1, 2 |
338 |
|
NR_134935.1 |
|
2, 3 |
391 |
|
NR_134936.1 |
|
2, 3 |
480 |
|
XM_011514619.2 |
XP_011512921.1
|
2, 3 |
477 |
|
XM_011514620.2 |
XP_011512922.1
|
2, 3 |
1264 |
|
XM_011514622.2 |
XP_011512924.1
|
2, 3 |
789 |
|
XM_017010863.1 |
XP_016866352.1
|
2, 3 |
790 |
|
XM_017010864.1 |
XP_016866353.1
|
2, 3 |
788 |
|
XM_017010866.1 |
XP_016866355.1
|
2, 3 |
443 |
|
GenBank mRNA |
AB593146.1 |
- |
2, 3 |
366 |
|
AF505623.1 |
- |
1, 2 |
338 |
|
AK097831.1 |
- |
2, 3 |
355 |
|
AK310329.1 |
- |
2, 3 |
355 |
|
AK314408.1 |
- |
2, 3 |
477 |
|
BC039825.1 |
- |
1, 2 |
345 |
5167621 |
BP246863.1 |
- |
2, 3 |
391 |
|
DB082719.1 |
- |
2, 3 |
379 |
|
JN226411.1 |
- |
1, 2 |
109 |
|
View Full Product Details