Gene Transcripts

Gene Symbol
SCN8A
Entrez Gene ID
6334
Gene Name
sodium voltage-gated channel alpha subunit 8
Gene Aliases
CERIII, CIAT, EIEE13, MED, NaCh6, Nav1.6, PN4
Chromosome Location
Chr. 12: 51589959 - 51812865 on Build GRCh38
UniGene ID
Hs.436550
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177984.2 NP_001171455.1 2 432
NM_014191.3 NP_055006.1 2 432
XM_006719556.3 XP_006719619.1 2 1023
XM_011538650.2 XP_011536952.1 2 451
XM_011538651.2 XP_011536953.1 2 416
XM_017019794.1 XP_016875283.1 2 429
XM_017019795.1 XP_016875284.1 2 448
XM_017019796.1 XP_016875285.1 2 448
GenBank mRNA AB027567.1 - 2 432
AF225988.1 - 1 254
AK309213.1 - 2 382
FJ611941.1 - 1 254

View Full Product Details