Gene Transcripts

Gene Symbol
PRKCH
Entrez Gene ID
5583
Gene Name
protein kinase C eta
Gene Aliases
PKC-L, PKCL, PRKCL, nPKC-eta
Chromosome Location
Chr. 14: 61280953 - 61550980 on Build GRCh38
UniGene ID
Hs.333907
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006255.4 NP_006246.2 14 2671
XM_011536954.2 XP_011535256.1 14 1854
XM_011536955.1 XP_011535257.1 14 1787
XM_017021458.1 XP_016876947.1 14 2005
GenBank mRNA AB451313.1 - 14 2012
AK093241.1 - 3 1564
AK290183.1 - 14 2390
AK296158.1 - 13 1685
AK300864.1 - 6 813
AX775931.1 - 14 1747
AX775933.1 - 14 2400
BC037268.1 - 14 2400 4822159
M55284.1 - 14 2175

View Full Product Details