Gene Transcripts

Gene Symbol
Chmp4c
Entrez Gene ID
66371
Gene Name
charged multivesicular body protein 4C
Gene Aliases
2010012P02Rik, 2210015K02Rik, 2310010I16Rik, Shax3, Snf7-3
Chromosome Location
Chr. 3: 10366973 - 10391005 on Build GRCm38
UniGene ID
Mm.366147
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025519.2 NP_079795.1 2 288
GenBank mRNA AK008217.1 - 1 1204
AK009273.1 - 2 288
AK010707.1 - 2 355
AK145033.1 - 2 292
AK160189.1 - 2 282
AK166371.1 - 2 310
BC115983.1 - 2 278 40105850
BC117538.1 - 2 278 40105849

View Full Product Details