Gene Transcripts

Gene Symbol
Fbxo5
Entrez Gene ID
67141
Gene Name
F-box protein 5
Gene Aliases
2510044I10Rik, C85305, Emi1, Fbxo31
Chromosome Location
Chr. 10: 5799158 - 5805465 on Build GRCm38
UniGene ID
Mm.197520
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025995.2 NP_080271.2 2 384
GenBank mRNA AK011820.1 - 2 382
AK135916.1 - 2 339
AK160201.1 - 2 340
BC053434.1 - 2 308 6336178

View Full Product Details