Gene Transcripts

Gene Symbol
1700067P10Rik
Entrez Gene ID
68224
Gene Name
RIKEN cDNA 1700067P10 gene
Gene Aliases
Chromosome Location
Chr. 17: 48089318 - 48090920 on Build GRCm38
UniGene ID
Mm.138490
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026625.2 NP_080901.2 3 332
GenBank mRNA AK015495.1 - 3 294
AK018887.1 - 3 321
BC048440.1 - 3 322 6741836

View Full Product Details