Gene Transcripts
- Gene Symbol
- Tmem180
- Entrez Gene ID
- 75146
- Gene Name
- transmembrane protein 180
- Gene Aliases
- 4930449A08Rik, 4930538D17Rik
- Chromosome Location
- Chr. 19: 46356877 - 46375254 on Build GRCm38
- UniGene ID
- Mm.159290
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_029186.2 |
NP_083462.2
|
5, 6 |
1016 |
|
GenBank mRNA |
AK015988.1 |
- |
2, 3 |
373 |
|
AK031554.1 |
- |
5, 6 |
962 |
|
AK052797.1 |
- |
2, 3 |
174 |
|
AK151797.1 |
- |
5, 6 |
991 |
|
AK161305.1 |
- |
5, 6 |
972 |
|
AK170247.1 |
- |
|
963 |
|
BC026385.1 |
- |
3, 4 |
1144 |
|
BC031169.1 |
- |
5, 6 |
961 |
|
BC049142.1 |
- |
2, 3 |
302 |
|
BC058746.1 |
- |
5, 6 |
962 |
6399588 |
View Full Product Details