Gene Transcripts

Gene Symbol
Card10
Entrez Gene ID
105844
Gene Name
caspase recruitment domain family, member 10
Gene Aliases
AI449026, Bimp1, Carma3
Chromosome Location
Chr. 15: 78775136 - 78803075 on Build GRCm38
UniGene ID
Mm.17629
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130859.2 NP_570929.2 4 867
GenBank mRNA AF363456.1 - 3 716
BC060203.1 - 4 867 6855024

View Full Product Details