Gene Transcripts
- Gene Symbol
- SPP1
- Entrez Gene ID
- 6696
- Gene Name
- secreted phosphoprotein 1
- Gene Aliases
- BNSP, BSPI, ETA-1, OPN
- Chromosome Location
- Chr. 4: 87975650 - 87983411 on Build GRCh38
- UniGene ID
- Hs.313
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000582.2 |
NP_000573.1
|
3, 4 |
257 |
|
NM_001040058.1 |
NP_001035147.1
|
3, 4 |
257 |
|
GenBank mRNA |
AF052124.1 |
- |
3, 4 |
179 |
|
AK057738.1 |
- |
3, 4 |
193 |
|
AK075463.1 |
- |
3, 4 |
193 |
|
AK290104.1 |
- |
3, 4 |
194 |
|
AK315461.1 |
- |
3, 4 |
187 |
|
AY956318.1 |
- |
3, 4 |
159 |
|
BC007016.1 |
- |
3, 4 |
193 |
4052438 |
BC017387.1 |
- |
3, 4 |
197 |
3828885 |
BC022844.1 |
- |
3, 4 |
194 |
4284921 |
BF699765.1 |
- |
3, 4 |
258 |
|
CB117856.1 |
- |
3, 4 |
257 |
|
D28759.1 |
- |
2, 3 |
92 |
|
D28760.1 |
- |
2, 3 |
92 |
|
DA419665.1 |
- |
3, 4 |
257 |
|
DQ839491.1 |
- |
2, 3 |
92 |
|
DQ846870.1 |
- |
2, 3 |
92 |
|
DQ846871.1 |
- |
2, 3 |
92 |
|
J04765.1 |
- |
3, 4 |
200 |
|
JF412666.1 |
- |
2, 3 |
94 |
|
M83248.1 |
- |
3, 4 |
189 |
|
X13694.1 |
- |
3, 4 |
159 |
|
View Full Product Details