Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133966.2 | NP_598727.1 | 3, 4 | 365 | |
GenBank mRNA | AK033477.1 | - | 3, 4 | 393 | |
AK089463.1 | - | 3, 4 | 445 | ||
AK154832.1 | - | 3, 4 | 445 | ||
BC013550.1 | - | 3, 4 | 365 | 4222283 |