Gene Transcripts
- Gene Symbol
- Spata20
- Entrez Gene ID
- 217116
- Gene Name
- spermatogenesis associated 20
- Gene Aliases
- Tisp78
- Chromosome Location
- Chr. 11: 94478904 - 94485310 on Build GRCm38
- UniGene ID
- Mm.156123
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_144827.4 |
NP_659076.2
|
16 |
2476 |
|
GenBank mRNA |
BC050788.1 |
- |
16 |
2476 |
|
View Full Product Details