Gene Transcripts

Gene Symbol
Nkx6-1
Entrez Gene ID
18096
Gene Name
NK6 homeobox 1
Gene Aliases
NKX6A, Nkx6.1
Chromosome Location
Chr. 5: 101659184 - 101664711 on Build GRCm38
UniGene ID
Mm.193072
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144955.2 NP_659204.1 3 1351
GenBank mRNA AF357883.1 - 3 1351
BC138019.1 - 3 1257 8861038
BC138020.1 - 3 1257 8861039

View Full Product Details