Gene Transcripts

Gene Symbol
Eva1a
Entrez Gene ID
232146
Gene Name
eva-1 homolog A (C. elegans)
Gene Aliases
BC014699, Fam176a, Tmem166
Chromosome Location
Chr. 6: 82041048 - 82093099 on Build GRCm38
UniGene ID
Mm.489683
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145570.2 NP_663545.2 3 1414
GenBank mRNA AK145140.1 - 3 1424
BC014699.1 - 3 1349 4221746
BC027150.1 - 3 1371 4950341

View Full Product Details