Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172447.2 | NP_766035.1 | 2, 3 | 384 | |
GenBank mRNA | AK029545.1 | - | 2, 3 | 385 | |
AK029553.1 | - | 2, 3 | 391 | ||
AK039313.1 | - | 2, 3 | 385 | ||
AK039388.1 | - | 2, 3 | 361 | ||
AK076835.1 | - | 2, 3 | 374 | ||
AK140211.1 | - | 2, 3 | 361 | ||
BC054079.1 | - | 2, 3 | 334 | ||
BC062906.1 | - | 2, 3 | 349 | 6813710 |