Gene Transcripts

Gene Symbol
Mfsd9
Entrez Gene ID
211798
Gene Name
major facilitator superfamily domain containing 9
Gene Aliases
4931419K03Rik
Chromosome Location
Chr. 1: 40772040 - 40790657 on Build GRCm38
UniGene ID
Mm.235443
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172499.2 NP_766087.1 6 755
GenBank mRNA AK029874.1 - 6 756
AK169911.1 - 7 916
BC057633.1 - 6 761 4035998

View Full Product Details