Gene Transcripts

Gene Symbol
Acad9
Entrez Gene ID
229211
Gene Name
acyl-Coenzyme A dehydrogenase family, member 9
Gene Aliases
2600017P15Rik, 4732402K02, C630012L17Rik, NPD002
Chromosome Location
Chr. 3: 36065984 - 36092857 on Build GRCm38
UniGene ID
Mm.260997
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172678.3 NP_766266.3 2 283
GenBank mRNA AK028571.1 - 2 211
AK031820.1 - 2 215
AK075984.1 - 2 202
AK145423.1 - 2 191
BC031137.1 - 2 219 4971848
BC032213.1 - 2 224 5323723
BC033277.1 - 2 204 4241041

View Full Product Details