Gene Transcripts

Gene Symbol
AI118078
Entrez Gene ID
244886
Gene Name
expressed sequence AI118078
Gene Aliases
9630029F15
Chromosome Location
Chr. 9: 55326919 - 55438345 on Build GRCm38
UniGene ID
Mm.24361
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172923.3 NP_766511.1 4 432
GenBank mRNA AK036040.1 - 4 432
BC068128.1 - 5 588 6835202

View Full Product Details